Assay Details
| Gene Symbol: FAM214B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353562.3 | XP_003353610.1 | 1412 | 75 | ||
| XM_003480544.4 | XP_003480592.1 | 1219 | 75 | |||
| XM_013986307.2 | XP_013841761.1 | 2676 | 75 | |||
| XM_013986310.2 | XP_013841764.1 | 1193 | 75 | |||
| XM_021065502.1 | XP_020921161.1 | 1546 | 75 | |||
More Information
Set Membership: |
|

FAM214B
Genomic Map