Assay Details
| Gene Symbol: ALAS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360337.4 | XP_003360385.1 | 1560 | 67 | ||
| XM_003360338.4 | XP_003360386.1 | 1449 | 67 | |||
| XM_005673669.3 | XP_005673726.1 | 1566 | 67 | |||
| GenBank mRNA | AK232940.1 | - | 5 - 6 | 1034 | 67 | |
| AK237661.1 | - | 8 - 9 | 1341 | 67 | ||
| AK396020.1 | - | 7 - 8 | 1187 | 67 | ||
More Information
Set Membership: |
|

ALAS2
Genomic Map