Assay Details
| Gene Symbol: CCDC92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005670575.3 | XP_005670632.1 | 717 | 66 | ||
| XM_005670576.3 | XP_005670633.1 | 1078 | 66 | |||
| XM_021072230.1 | XP_020927889.1 | 874 | 66 | |||
| XM_021072231.1 | XP_020927890.1 | 868 | 66 | |||
| XM_021072232.1 | XP_020927891.1 | 572 | 66 | |||
| XM_021072233.1 | XP_020927892.1 | 369 | 66 | |||
| XM_021072234.1 | XP_020927893.1 | 751 | 66 | |||
| XM_021072235.1 | XP_020927894.1 | 393 | 66 | |||
| XM_021072236.1 | XP_020927895.1 | 1403 | 66 | |||
| GenBank mRNA | AK233129.1 | - | 2 - 3 | 369 | 66 | |
More Information
Set Membership: |
|

CCDC92
Genomic Map