Assay Details
| Gene Symbol: DNAJC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355378.4 | XP_003355426.1 | 1236 | 91 | ||
| XM_005655559.3 | XP_005655616.1 | 1384 | 91 | |||
| XM_021091601.1 | XP_020947260.1 | 1236 | 91 | |||
| GenBank mRNA | AK233192.1 | - | 2 - 3 | 1051 | 91 | |
| AK392918.1 | - | 2 - 3 | 1068 | 91 | ||
| AK394334.1 | - | 2 - 3 | 1051 | 91 | ||
More Information
Set Membership: |
|

DNAJC22
Genomic Map