Assay Details
| Gene Symbol: MRPL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123056.5 | XP_003123104.1 | 324 | 93 | ||
| GenBank mRNA | AK233221.1 | - | 2 - 3 | 310 | 93 | |
| AK395605.1 | - | 2 - 3 | 313 | 93 | ||
More Information
Set Membership: |
|

MRPL54
Genomic Map