Assay Details
| Gene Symbol: GUCY1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130093.6 | XP_003130141.3 | 918 | 78 | ||
| GenBank mRNA | AK233223.1 | - | 4 - 5 | 388 | 78 | |
| AK345330.1 | - | 3 - 4 | 883 | 78 | ||
More Information
Set Membership: |
|

GUCY1A2
Genomic Map