Assay Details
| Gene Symbol: MRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013993955.2 | XP_013849409.1 | 231 | 95 | ||
| GenBank mRNA | AK233229.1 | - | 199 | 95 | ||
| AK352439.1 | - | 178 | 95 | |||
| AK397977.1 | - | 236 | 95 | |||
More Information
Set Membership: |
|

MRPS2
Genomic Map