Assay Details
Gene Symbol: TEX264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068857.1 | XP_020924516.1 | 867 | 70 | ||
XM_021068858.1 | XP_020924517.1 | 982 | 70 | |||
XM_021068859.1 | XP_020924518.1 | 1659 | 70 | |||
XM_021068861.1 | XP_020924520.1 | 913 | 70 | |||
XM_021068862.1 | XP_020924521.1 | 988 | 70 | |||
XM_021068863.1 | XP_020924522.1 | 914 | 70 | |||
XM_021068864.1 | XP_020924523.1 | 646 | 70 | |||
GenBank mRNA | AK233248.1 | - | 5 - 6 | 797 | 70 | |
AK346217.1 | - | 5 - 6 | 797 | 70 | ||
AK389349.1 | - | 5 - 6 | 823 | 70 | ||
AK390206.1 | - | 5 - 6 | 987 | 70 | ||
AK400465.1 | - | 4 - 5 | 1093 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
