Assay Details
| Gene Symbol: DEPTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090553.1 | XP_020946212.1 | 1878 | 99 | ||
| XM_021090554.1 | XP_020946213.1 | 1878 | 99 | |||
| XM_021090555.1 | XP_020946214.1 | 1835 | 99 | |||
| XM_021090556.1 | XP_020946215.1 | 1878 | 99 | |||
| XM_021090557.1 | XP_020946216.1 | 1878 | 99 | |||
| XM_021090558.1 | XP_020946217.1 | 1878 | 99 | |||
| GenBank mRNA | AK233254.1 | - | 6 - 7 | 1014 | 99 | |
| AK394604.1 | - | 4 - 5 | 602 | 99 | ||
More Information
Set Membership: |
|

DEPTOR
Genomic Map