Assay Details
| Gene Symbol: TMEM53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482099.4 | XP_003482147.1 | 284 | 102 | ||
| XM_021096785.1 | XP_020952444.1 | 164 | 102 | |||
| GenBank mRNA | AK233358.1 | - | 1 - 2 | 108 | 102 | |
| AK396847.1 | - | 1 - 2 | 122 | 102 | ||
More Information
Set Membership: |
|

TMEM53
Genomic Map