Assay Details
| Gene Symbol: CHST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132414.6 | XP_003132462.2 | 430 | 102 | ||
| GenBank mRNA | AK233433.1 | - | 1 - 2 | 141 | 102 | |
More Information
Set Membership: |
|

CHST13
Genomic Map