Assay Details
| Gene Symbol: CHST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132414.6 | XP_003132462.2 | 504 | 93 | ||
| GenBank mRNA | AK233433.1 | - | 2 - 3 | 215 | 93 | |
More Information
Set Membership: |
|

CHST13
Genomic Map