Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099141.1 | XP_020954800.1 | 3392 | 64 | ||
| XM_021099142.1 | XP_020954801.1 | 3531 | 64 | |||
| XM_021099143.1 | XP_020954802.1 | 3534 | 64 | |||
| XM_021099144.1 | XP_020954803.1 | 4133 | 64 | |||
| XM_021099145.1 | XP_020954804.1 | 3390 | 64 | |||
| XM_021099146.1 | XP_020954805.1 | 3390 | 64 | |||
| XM_021099148.1 | XP_020954807.1 | 3387 | 64 | |||
| GenBank mRNA | AK233475.1 | - | 5 - 6 | 719 | 64 | |
| AK345811.1 | - | 5 - 6 | 688 | 64 | ||
More Information
Set Membership: |
|

SLC25A21
Genomic Map