Assay Details
| Gene Symbol: CCDC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063068.1 | XP_020918727.1 | 2211 | 64 | ||
| XM_021063069.1 | XP_020918728.1 | 2530 | 64 | |||
| XM_021063070.1 | XP_020918729.1 | 2051 | 64 | |||
| XM_021063071.1 | XP_020918730.1 | 1879 | 64 | |||
| XM_021063072.1 | XP_020918731.1 | 1844 | 64 | |||
| XM_021063073.1 | XP_020918732.1 | 2090 | 64 | |||
| XM_021063074.1 | XP_020918733.1 | 2087 | 64 | |||
| XM_021063075.1 | XP_020918734.1 | 2406 | 64 | |||
| XM_021063077.1 | XP_020918736.1 | 1755 | 64 | |||
| XM_021063078.1 | XP_020918737.1 | 1721 | 64 | |||
| XM_021063079.1 | XP_020918738.1 | 1966 | 64 | |||
| XM_021063080.1 | XP_020918739.1 | 1635 | 64 | |||
| XM_021063081.1 | XP_020918740.1 | 1600 | 64 | |||
| XM_021063082.1 | XP_020918741.1 | 1527 | 64 | |||
| XM_021063083.1 | XP_020918742.1 | 2212 | 64 | |||
| XM_021063084.1 | XP_020918743.1 | 2212 | 64 | |||
More Information
Set Membership: |
|

CCDC15
Genomic Map