Assay Details
| Gene Symbol: CCDC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066369.1 | XP_020922028.1 | 2791 | 53 | ||
| XM_021066370.1 | XP_020922029.1 | 2847 | 53 | |||
More Information
Set Membership: |
|

CCDC57
Genomic Map