Assay Details
| Gene Symbol: ROR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097759.1 | XP_020953418.1 | 2232 | 100 | ||
| XM_021097760.1 | XP_020953419.1 | 2219 | 100 | |||
| XM_021097761.1 | XP_020953420.1 | 2075 | 100 | |||
| XM_021097763.1 | XP_020953422.1 | 1793 | 100 | |||
| XM_021097764.1 | XP_020953423.1 | 1638 | 100 | |||
| XM_021097765.1 | XP_020953424.1 | 2236 | 100 | |||
| XM_021097766.1 | XP_020953425.1 | 2236 | 100 | |||
More Information
Set Membership: |
|

ROR1
Genomic Map