Assay Details
| Gene Symbol: AMIGO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663589.3 | XP_005663646.1 | 351 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005663589.3 transcripts.
Set Membership: |
|

AMIGO1
Genomic Map