Assay Details
| Gene Symbol: NHLH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089573.1 | XP_020945232.1 | 3036 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021089573.1 transcripts.
Set Membership: |
|

NHLH1
Genomic Map