Assay Details
| Gene Symbol: SLC41A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098938.1 | XP_020954597.1 | 966 | 60 | ||
| XM_021098940.1 | XP_020954599.1 | 1098 | 60 | |||
| XM_021098941.1 | XP_020954600.1 | 874 | 60 | |||
| XM_021098942.1 | XP_020954601.1 | 982 | 60 | |||
| XM_021098943.1 | XP_020954602.1 | 1093 | 60 | |||
| XM_021098944.1 | XP_020954603.1 | 684 | 60 | |||
More Information
Set Membership: |
|

SLC41A3
Genomic Map