Assay Details
| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132489.3 | XP_003132537.1 | 463 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003132489.3 transcripts.
Set Membership: |
|

GPR87
Genomic Map