Assay Details
| Gene Symbol: B4GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096829.1 | XP_020952488.1 | 879 | 57 | ||
| XM_021096830.1 | XP_020952489.1 | 880 | 57 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4GALT2| Gene Symbol: B4GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096829.1 | XP_020952488.1 | 879 | 57 | ||
| XM_021096830.1 | XP_020952489.1 | 880 | 57 | |||
Set Membership: |
|