Assay Details
| Gene Symbol: RAPSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122810.6 | XP_003122858.5 | 864 | 67 | ||
| XM_013994350.2 | XP_013849804.2 | 1805 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RAPSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122810.6 | XP_003122858.5 | 864 | 67 | ||
| XM_013994350.2 | XP_013849804.2 | 1805 | 67 | |||
Set Membership: |
|