Assay Details
| Gene Symbol: FUT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081782.1 | XP_020937441.1 | 852 | 78 | ||
| XM_021081783.1 | XP_020937442.1 | 852 | 78 | |||
| XM_021081784.1 | XP_020937443.1 | 852 | 78 | |||
| XM_021081785.1 | XP_020937444.1 | 852 | 78 | |||
| XM_021081786.1 | XP_020937445.1 | 852 | 78 | |||
| XM_021081787.1 | XP_020937446.1 | 852 | 78 | |||
| XM_021081788.1 | XP_020937447.1 | 852 | 78 | |||
| XM_021081790.1 | XP_020937449.1 | 502 | 78 | |||
More Information
Set Membership: |
|

FUT10
Genomic Map