Assay Details
| Gene Symbol: SLC7A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128550.6 | XP_003128598.5 | 817 | 61 | ||
| XM_021099239.1 | XP_020954898.1 | 1863 | 61 | |||
| XM_021099240.1 | XP_020954899.1 | 1281 | 61 | |||
More Information
Set Membership: |
|

SLC7A8
Genomic Map