Assay Details
| Gene Symbol: SYPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125869.4 | XP_003125917.1 | 712 | 82 | ||
| XM_021090083.1 | XP_020945742.1 | 714 | 82 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SYPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125869.4 | XP_003125917.1 | 712 | 82 | ||
| XM_021090083.1 | XP_020945742.1 | 714 | 82 | |||
Set Membership: |
|