Assay Details
| Gene Symbol: GPR63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013992707.2 | XP_013848161.1 | 799 | 149 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013992707.2 transcripts.
Set Membership: |
|

GPR63
Genomic Map