Assay Details
| Gene Symbol: LOC100155738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131007.5 | XP_003131055.2 | 4126 | 78 | ||
| XM_013980572.2 | XP_013836026.2 | 4417 | 78 | |||
More Information
Set Membership: |
|

LOC100155738
Genomic Map