Assay Details
| Gene Symbol: KCTD16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084950.1 | XP_020940609.1 | 1245 | 85 | ||
| XM_021084951.1 | XP_020940610.1 | 970 | 85 | |||
| XM_021084952.1 | XP_020940611.1 | 844 | 85 | |||
| XM_021084953.1 | XP_020940612.1 | 803 | 85 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021084950.1, XM_021084951.1, XM_021084952.1, XM_021084953.1 transcripts.
Set Membership: |
|

KCTD16
Genomic Map