Assay Details
| Gene Symbol: MAGEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098674.1 | XP_020954333.1 | 2282 | 65 | ||
| GenBank mRNA | GU117604.1 | - | 1 | 519 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MAGEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098674.1 | XP_020954333.1 | 2282 | 65 | ||
| GenBank mRNA | GU117604.1 | - | 1 | 519 | 65 | |
Set Membership: |
|