Assay Details
| Gene Symbol: IAPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126437.4 | XP_003126485.2 | 405 | 82 | ||
| GenBank mRNA | GU396090.1 | - | 1 | 48 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IAPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126437.4 | XP_003126485.2 | 405 | 82 | ||
| GenBank mRNA | GU396090.1 | - | 1 | 48 | 82 | |
Set Membership: |
|