Assay Details
| Gene Symbol: ANGPTL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083871.1 | XP_020939530.1 | 1505 | 117 | ||
| XM_021083872.1 | XP_020939531.1 | 1495 | 117 | |||
| GenBank mRNA | KF017598.1 | - | 2 - 3 | 462 | 117 | |
| KX447621.1 | - | 2 - 3 | 480 | 117 | ||
More Information
Set Membership: |
|

ANGPTL8
Genomic Map