Assay Details
| Gene Symbol: RNASEH2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660692.2 | XP_005660749.1 | 433 | 70 | ||
| GenBank mRNA | AK233803.1 | - | 2 - 3 | 391 | 70 | |
| AK399704.1 | - | 2 - 3 | 404 | 70 | ||
More Information
Set Membership: |
|

RNASEH2C
Genomic Map