Assay Details
| Gene Symbol: NMI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359414.4 | XP_003359462.2 | 602 | 106 | ||
| GenBank mRNA | AK234080.1 | - | 4 - 5 | 571 | 106 | |
| AK351870.1 | - | 4 - 5 | 533 | 106 | ||
| AK393878.1 | - | 5 - 6 | 508 | 106 | ||
More Information
Set Membership: |
|

NMI
Genomic Map