Assay Details
| Gene Symbol: ALG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358712.4 | XP_003358760.1 | 1053 | 72 | ||
| XM_005670035.3 | XP_005670092.2 | 1375 | 72 | |||
| GenBank mRNA | AK234203.1 | - | 7 - 8 | 1015 | 72 | |
More Information
Set Membership: |
|

ALG3
Genomic Map