Assay Details
| Gene Symbol: ADPGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021099099.1 | XP_020954758.1 | 597 | 70 | ||
| XM_021099100.1 | XP_020954759.1 | 599 | 70 | |||
| GenBank mRNA | AK234411.1 | - | 3 - 4 | 581 | 70 | |
More Information
Set Membership: |
|

ADPGK
Genomic Map