Assay Details
| Gene Symbol: ETV6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924598.6 | XP_001924633.3 | 1442 | 73 | ||
| XM_005652587.3 | XP_005652644.1 | 1440 | 73 | |||
| GenBank mRNA | AK234911.1 | - | 7 - 8 | 1371 | 73 | |
| AK398062.1 | - | 7 - 8 | 1401 | 73 | ||
More Information
Set Membership: |
|

ETV6
Genomic Map