Assay Details
| Gene Symbol: ZNF827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101535.1 | XP_020957194.1 | 1653 | 76 | ||
| XM_021101536.1 | XP_020957195.1 | 1659 | 76 | |||
| XM_021101537.1 | XP_020957196.1 | 1652 | 76 | |||
| XM_021101538.1 | XP_020957197.1 | 1652 | 76 | |||
| XM_021101539.1 | XP_020957198.1 | 1651 | 76 | |||
| XM_021101540.1 | XP_020957199.1 | 1651 | 76 | |||
| XM_021101541.1 | XP_020957200.1 | 1651 | 76 | |||
| GenBank mRNA | AK235100.1 | - | 2 - 3 | 1128 | 76 | |
More Information
Set Membership: |
|

ZNF827
Genomic Map