Assay Details
| Gene Symbol: MED4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013978727.2 | XP_013834181.1 | 169 | 101 | ||
| GenBank mRNA | AK235167.1 | - | 1 - 2 | 138 | 101 | |
| AK238920.1 | - | 1 - 2 | 138 | 101 | ||
| AK239579.1 | - | 1 - 2 | 139 | 101 | ||
More Information
Set Membership: |
|

MED4
Genomic Map