Assay Details
| Gene Symbol: RNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005665930.3 | XP_005665987.2 | 1330 | 71 | ||
| XM_005665931.3 | XP_005665988.1 | 1305 | 71 | |||
| XM_005665932.3 | XP_005665989.2 | 1340 | 71 | |||
| XM_013977872.2 | XP_013833326.2 | 1304 | 71 | |||
| XM_013977873.2 | XP_013833327.2 | 1179 | 71 | |||
| XM_021098656.1 | XP_020954315.1 | 1136 | 71 | |||
| GenBank mRNA | AK235343.1 | - | 6 - 7 | 1342 | 71 | |
| AK238751.1 | - | 5 - 6 | 1266 | 71 | ||
| AK390287.1 | - | 5 - 6 | 1296 | 71 | ||
More Information
Set Membership: |
|

RNF8
Genomic Map