Assay Details
| Gene Symbol: GNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003480989.3 | XP_003481037.1 | 1169 | 70 | ||
| XM_021086249.1 | XP_020941908.1 | 1009 | 70 | |||
| GenBank mRNA | AK235450.1 | - | 9 - 10 | 1115 | 70 | |
| AK391974.1 | - | 9 - 10 | 1170 | 70 | ||
More Information
Set Membership: |
|

GNB2
Genomic Map