Assay Details
| Gene Symbol: SETD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656533.3 | XP_005656590.2 | 172 | 65 | ||
| XM_021101559.1 | XP_020957218.1 | 527 | 65 | |||
| XM_021101560.1 | XP_020957219.1 | 457 | 65 | |||
| GenBank mRNA | AK235540.1 | - | 1 - 2 | 115 | 65 | |
More Information
Set Membership: |
|

SETD7
Genomic Map