Assay Details
| Gene Symbol: PPM1M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132254.6 | XP_003132302.3 | 890 | 59 | ||
| XM_013981765.2 | XP_013837219.1 | 1092 | 59 | |||
| GenBank mRNA | AK235558.1 | - | 3 - 4 | 444 | 59 | |
| AK346848.1 | - | 1 - 2 | 94 | 59 | ||
| AK390760.1 | - | 4 - 5 | 506 | 59 | ||
More Information
Set Membership: |
|

PPM1M
Genomic Map