Assay Details
| Gene Symbol: JMJD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066561.1 | XP_020922220.1 | 787 | 73 | ||
| GenBank mRNA | AK235664.1 | - | 2 - 3 | 649 | 73 | |
| AK347723.1 | - | 2 - 3 | 620 | 73 | ||
| AK389326.1 | - | 2 - 3 | 683 | 73 | ||
More Information
Set Membership: |
|

JMJD6
Genomic Map