Assay Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926616.6 | XP_001926651.2 | 465 | 85 | ||
| GenBank mRNA | AK235675.1 | - | 3 - 4 | 363 | 86 | |
| AK235951.1 | - | 3 - 4 | 402 | 85 | ||
| AK393130.1 | - | 3 - 4 | 414 | 85 | ||
| AK400182.1 | - | 3 - 4 | 423 | 86 | ||
| AY609427.1 | - | 3 - 4 | 395 | 86 | ||
More Information
Set Membership: |
|

ZC3H15
Genomic Map