Assay Details
| Gene Symbol: SNTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093874.1 | XP_020949533.1 | 636 | 68 | ||
| GenBank mRNA | AK235680.1 | - | 1 - 2 | 585 | 68 | |
| AK346504.1 | - | 1 - 2 | 611 | 68 | ||
| AK348139.1 | - | 1 - 2 | 585 | 68 | ||
More Information
Set Membership: |
|

SNTB2
Genomic Map