Assay Details
| Gene Symbol: MPZL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663141.3 | XP_005663198.1 | 456 | 80 | ||
| XM_013996806.2 | XP_013852260.1 | 458 | 80 | |||
| GenBank mRNA | AK235730.1 | - | 2 - 3 | 380 | 80 | |
| AK237891.1 | - | 2 - 3 | 376 | 80 | ||
| AK395406.1 | - | 2 - 3 | 450 | 80 | ||
More Information
Set Membership: |
|

MPZL1
Genomic Map