Assay Details
| Gene Symbol: IL11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095008.1 | XP_020950667.1 | 351 | 116 | ||
| GenBank mRNA | AB198667.1 | - | 1 - 2 | 181 | 116 | |
More Information
Set Membership: |
|

IL11
Genomic Map