Assay Details
| Gene Symbol: MTERF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130132.4 | XP_003130180.1 | 123 | 104 | ||
| GenBank mRNA | AK230494.1 | - | 2 - 3 | 105 | 104 | |
| AK389868.1 | - | 2 - 3 | 112 | 104 | ||
More Information
Set Membership: |
|

MTERF1
Genomic Map