Assay Details
| Gene Symbol: VPS4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121699.5 | XP_003121747.3 | 397 | 75 | ||
| GenBank mRNA | AK230524.1 | - | 3 - 4 | 325 | 75 | |
| AK389854.1 | - | 2 - 3 | 364 | 75 | ||
| AK395399.1 | - | 2 - 3 | 367 | 75 | ||
More Information
Set Membership: |
|

VPS4B
Genomic Map