Assay Details
| Gene Symbol: PSMA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003360207.5 | XP_003360255.2 | 208 | 58 | ||
| GenBank mRNA | AK230619.1 | - | 2 - 3 | 123 | 58 | |
| AK343849.1 | - | 2 - 3 | 132 | 58 | ||
| AK343855.1 | - | 2 - 3 | 132 | 58 | ||
| AK343887.1 | - | 2 - 3 | 132 | 58 | ||
| AK346795.1 | - | 2 - 3 | 138 | 58 | ||
More Information
Set Membership: |
|

PSMA2
Genomic Map